missing translation for 'onlineSavingsMsg'
Learn More

Esculin Monohydrate, ≥97.5%, MP Biomedicals™

Product Code. 11478020 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478020 100 g 100g
11458020 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478020 Supplier MP Biomedicals Supplier No. 0210165580
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Molecular Biology

Chemical Identifiers

CAS 531-75-9
Molecular Formula C15H16O9
Molecular Weight (g/mol) 340.28
MDL Number MFCD00149492
InChI Key XHCADAYNFIFUHF-TYKRLAFXNA-N
Synonym esculin, aesculin, esculoside, --esculin, polychrome, 6,7-dihydroxycoumarin-6-o-glucoside, aesculinum, esculine, esculetin 6-o-glucoside, 6,7-dihydroxycoumarin 6-glucoside
PubChem CID 5281417
ChEBI CHEBI:4853
IUPAC Name 7-hydroxy-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one
SMILES OC[C@H]1O[C@@H](OC2=C(O)C=C3OC(=O)C=CC3=C2)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Assay Percent Range ≥97.5%
Quantity 100 g
Formula Weight 358.3
Physical Form Solid
Chemical Name or Material Esculin Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.