missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ESCO2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA524210

Product Code. 17146029

  • £478.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ESCO2 Polyclonal Antibody for Western Blot, Flow

This gene encodes a protein that may have acetyltransferase activity and may be required for the establishment of sister chromatid cohesion during the S phase of mitosis. Mutations in this gene have been associated with Roberts syndrome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ESCO2
Polyclonal
Unconjugated
ESCO2
2410004I17Rik; D030072L07Rik; ECO1 homolog 2; EFO2; Esco2; Establishment factor-like protein 2; Establishment of cohesion 1 homolog 2; establishment of cohesion 1 homolog 2 (S. cerevisiae); establishment of sister chromatid cohesion N-acetyltransferase 2; N-acetyltransferase ESCO2; RBS
Rabbit
Antigen affinity chromatography, Protein A
RUO
157570, 71988
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Western Blot
2 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q56NI9, Q8CIB9
ESCO2
KLH conjugated synthetic peptide between 132-161 amino acids from the central region of human ESCO2.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.