Learn More
Invitrogen™ ESCO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113883
Description
Immunogen sequence: TEVSRRSLRS REIQGQVQAV KQSLPPTKKE QCSSTQSKSN KTSQKHVKRK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 79%; Dog: 85%; Horse: 81%; Human: 100%; Pig: 75%; Rat: 85%.
EFO1 (Establishment factor-like protein 1), also known as ESCO1 (establishment of cohesion 1 homolog1) or ESO1, is a member of the acetyltransferase family (GCN5 subfamily). It is a ubiquitously expressed nuclear protein that plays an important role in sister chromatid cohesion. At its C-terminus, EFO1 contains an H2C2 zinc finger motif and an acetyltransferase domain that exhibits acetyltransferase activity in vivo. Its N-terminus, containing two domains that are similar o alpha- and beta-type linker histone proteins, is essential for EFO1 association with chromosomes. EFO1 is responsible for coupling cohesion and DNA replication processes thereby ensuring proper pairing of sister chromatids. EFO1 is phosphorylated during mitosis and this may act to regulate EFO1 activity. Due to alternative splicing events, three EFO1 isoforms exist.
Specifications
| ESCO1 | |
| Polyclonal | |
| Unconjugated | |
| ESCO1 | |
| A930014I12Rik; CTF; CTF7 homolog 1; ECO1; ECO1 homolog 1; EFO1; EFO1p; ESCO1; ESO1; ESO1 homolog 1; establishment factor-like protein 1; Establishment of cohesion 1 homolog 1; establishment of cohesion 1 homolog 1 (S. cerevisiae); establishment of sister chromatid cohesion N-acetyltransferase 1; establisment of cohesion 1 homolog 1; hEFO1; Kiaa1911; N-acetyltransferase ESCO1; N-acetyltransferase ESCO1 variant 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114799 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5FWF5 | |
| ESCO1 | |
| Immunogen sequence: TEVSRRSLRS REIQGQVQAV KQSLPPTKKE QCSSTQSKSN KTSQKHVKRK (aa 129-178). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.