missing translation for 'onlineSavingsMsg'
Learn More

Escitalopram oxalate, Tocris Bioscience™

Product Code. 30001558
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective serotonin reuptake inhibitor (SSRI)

Escitalopram oxalate is a selective 5-HT reuptake inhibitor (Ki values are 0.89, 8150, and 10500 nM for serotonin, noradrenalin, and dopamine transporters respectively). Escitalopram oxalate is an S-enantiomer and eutomer of citalopram (Cat. No. 1427).

Chemical Identifiers

CAS 219861-08-2
Molecular Formula C22H23FN2O5
Molecular Weight (g/mol) 414.433
InChI Key KTGRHKOEFSJQNS-BDQAORGHSA-N
Synonym escitalopram oxalate, lexapro, cipralex, s-citalopram oxalate, citalopram oxalate, unii-5u85dbw7lo, 5u85dbw7lo, s-+-citalopram oxalate, s-1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydroisobenzofuran-5-carbonitrile oxalate
PubChem CID 146571
IUPAC Name (1S)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;oxalic acid
SMILES CN(C)CCCC1(C2=C(CO1)C=C(C=C2)C#N)C3=CC=C(C=C3)F.C(=O)(C(=O)O)O

Specifications

Quantity 50 mg
Formula Weight 414.43
Percent Purity 98%
Chemical Name or Material Escitalopram oxalate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'