missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Escitalopram Oxalate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Escitalopram Oxalate
Specifications
Specifications
| Chemical Name or Material | Escitalopram Oxalate |
| CAS | 219861-08-2 |
| Analyte or Component Names | Escitalopram Oxalate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H21 F N2 O . C2 H2 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Escitalopram Oxalate, (S)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile Ethanedioate, (+)-(S)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile Oxalate |
| InChI Formula | InChI=1S/C20H21FN2O.C2H2O4/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20;3-1(4)2(5)6/h4-9,12H,3,10-11,14H2,1-2H3;(H,3,4)(H,5,6)/t20-;/m0./s1 |
| SMILES | CN(C)CCC[C@]1(OCc2cc(ccc12)C#N)c3ccc(F)cc3.OC(=O)C(=O)O |
| IUPAC Name | (1S)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;oxalic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction