missing translation for 'onlineSavingsMsg'
Learn More

Escin, MedChemExpress

Product Code. 16428738
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16428738 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16428738 Supplier Medchem Express Supplier No. HYB2114

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Escin, a natural compound of triterpenoid saponins isolated from horse chestnut (Aesculus hippocastanum) seeds, can be used as a vasoprotective anti-inflammatory, anti-edematous and anti-nociceptive agent.

Escin, a natural compound of triterpenoid saponins isolated from horse chestnut (Aesculus hippocastanum) seeds, can be used as a vasoprotective anti-inflammatory, anti-edematous and anti-nociceptive agent.

Specifications

Chemical Name or Material Escin
CAS 6805-41-0
Color Light Nude
Purity Grade Notes Research
Molecular Formula C55H86O24
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (88.40 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1(C)[C@@H](OC(/C(C)=C/C)=O)[C@H](OC(C)=O)[C@@]([C@H](O)C[C@]2(C)C3=CC[C@@]4([H])[C@@]2(C)CC[C@]5([H])[C@]4(C)CC[C@H](O[C@@H]6O[C@H](C(O)=O)[C@@H](O[C@@H]7[C@@H](O)[C@H](O)[C@@H](O)[C@H](CO)O7)[C@H](O)[C@H]6O[C@@H]8OC(CO)[C@@H](O)[C@H](O)[C@H]8O)[C@@]5(CO)C)(CO)[C@@]3([H])C1
Molecular Weight (g/mol) 1131.26
Formula Weight 1131.26
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.