missing translation for 'onlineSavingsMsg'
Learn More

β-Escin, 92 to 95%, MP Biomedicals™

Product Code. 11425381 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g

Product Code. 11425381

Brand: MP Biomedicals 0215794101

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 11072-93-8
Molecular Formula C110H172O48
Molecular Weight (g/mol) 2262.54
MDL Number MFCD00076054
InChI Key YFESOSRPNPYODN-LNXBMLMBSA-N
Synonym aescin, flogencyl, beta escin, 2s,3s,4s,5r,6r-6-3s,4s,6ar,6bs,8r,8ar,9r,10r,14br-9-acetyloxy-8-hydroxy-4,8a-bis hydroxymethyl-4,6a,6b,11,11,14b-hexamethyl-10-2z-2-methylbut-2-enoyl oxy-1,2,3,4a,5,6,7,8,9,10,12,12a,14,14a-tetradecahydropicen-3-yl oxy-4-hydroxy-3,5-bis 2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy oxane-2-carboxylic acid
PubChem CID 16211024
IUPAC Name (2S,3S,4S,5R,6R)-6-{[(3S,4S,4aR,6aR,6bS,8R,8aR,9R,10R,12aS,14aR,14bR)-9-(acetyloxy)-8-hydroxy-4,8a-bis(hydroxymethyl)-4,6a,6b,11,11,14b-hexamethyl-10-{[(2Z)-2-methylbut-2-enoyl]oxy}-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,14,14a,14b-icosahydropicen-3-yl]oxy}-4-hydroxy-3,5-bis({[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})oxane-2-carboxylic acid; (2S,3S,4S,5R,6R)-6-{[(3S,4S,6aR,6bS,8R,8aR,9R,10R,14bR)-9-(acetyloxy)-8-hydroxy-4,8a-bis(hydroxymethyl)-4,6a,6b,11,11,14b-hexamethyl-10-{[(2E)-2-methylbut-2-enoyl]oxy}-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,14,14a,14b-icosahydropicen-3-yl]oxy}-4-hydroxy-3,5-bis({[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})oxane-2-carboxylic acid
SMILES C\C=C(\C)C(=O)O[C@H]1[C@H](OC(C)=O)[C@]2(CO)[C@H](O)C[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O[C@@H]6O[C@@H]([C@@H](O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)[C@H](O)[C@H]6O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)C(O)=O)[C@](C)(CO)[C@@H]5CC[C@@]34C)[C@@H]2CC1(C)C.C\C=C(/C)C(=O)O[C@H]1[C@H](OC(C)=O)[C@]2(CO)[C@H](O)C[C@]3(C)C(=CCC4[C@@]5(C)CC[C@H](O[C@@H]6O[C@@H]([C@@H](O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)[C@H](O)[C@H]6O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)C(O)=O)[C@](C)(CO)C5CC[C@@]34C)C2CC1(C)C

Specifications

Color White
Assay Percent Range 92 to 95%
Quantity 1 g
Formula Weight 1101.2
Physical Form Powder
Chemical Name or Material β-Escin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.