missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESAM Antibody (113), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89321IR
This item is not returnable.
View return policy
Description
ESAM Monoclonal specifically detects ESAM in Human samples. It is validated for Flow Cytometry.
Specifications
| ESAM | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 90952 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry | |
| 113 | |
| Flow Cytometry | |
| 2310008D05Rik, endothelial cell adhesion molecule, endothelial cell-selective adhesion molecule, HUEL (C4orf1)-interacting protein, LP4791 protein, W117m | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ESAM (NP_620411.2; Met1-Ala248). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction