missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ ES 7X™-PF Cleaning Solution
GREENER_CHOICE

Product Code. 17104185 Shop All MP Biomedicals Products
Click to view available options
Volume (English):
1 gal.
1.32
4 x 1 gal.
5 gal.
55 gal.
Unit Size:
10 L
18.92 L
208.19 L
Each
Pack of 4
Pack of 5
Pack of 55
This item is not returnable. View return policy

Product Code. 17104185

Brand: MP Biomedicals™ 097667195

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ES 7X™-PF Cleaning Solution is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.

  • Environmentally safe and phosphate free.
  • Developed to provide safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
  • Several convenient package sizes are available
  • Sold in concentrated form so the customer can prepare the appropriate dilution as needed
  • Completely drains from glassware in seconds
  • Eliminates residue leaving no interfering fluorescence
  • pH neutral, and no need for pH adjustment at any concentration
  • No etch to glass or plastic labware at any concentration
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
pH 7 to 8
Appearance Yellow
Concentration 7X
Content And Storage 15°C to 30°C
For Use With (Application) Use on Glass, Plastic, benchtop equipment and tubing
Volume (English) 5 gal.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.