missing translation for 'onlineSavingsMsg'
Learn More

Erythrosin B, spirit soluble

Product Code. 15474949
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15474949

Brand: Thermo Scientific Alfa Aesar J62619.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 15905-32-5 | C20H8I4O5 | 835.90 g/mol

Erythrosin B is used as a red coloring in some foods such as cherries and fish. It is used as printing ink, dental plaque disclosing agent and biological stain. It finds application as a radiopaque medium and sensitizer for orthochromatic photographic films. It is utilized as a cake-decorating gel. In addition to this, it is used in sweets such as candies and popsicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Erythrosin B is used as a red coloring in some foods such as cherries and fish. It is used as printing ink, dental plaque disclosing agent and biological stain. It finds application as a radiopaque medium and sensitizer for orthochromatic photographic films. It is utilized as a cake-decorating gel. In addition to this, it is used in sweets such as candies and popsicles.

Solubility
Soluble in water, spirit, methanol, methyl cellosolve and ethanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15905-32-5
Molecular Formula C20H8I4O5
Molecular Weight (g/mol) 835.90
MDL Number MFCD00005044
InChI Key OALHHIHQOFIMEF-UHFFFAOYSA-N
Synonym C.I. 45430:2; Solvent Red 140
PubChem CID 3259
SMILES OC1=C(I)C2=C(C=C1I)C1(OC(=O)C3=C1C=CC=C3)C1=CC(I)=C(O)C(I)=C1O2

Specifications

Solubility Information Soluble in water,spirit,methanol,methyl cellosolve and ethanol.
IUPAC Name 3',6'-dihydroxy-2',4',5',7'-tetraiodo-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Formula Weight 835.89
Color Orange to Orange/Red
Quantity 5 g
Physical Form Powder
Chemical Name or Material Erythrosin B
compliance-icons
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.