missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erythrosin B, ∽98%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| CAS | 15905-32-5 |
| Assay Percent Range | ∽98% |
| Molecular Formula | C20H8I4O5 |
| MDL Number | MFCD00005044 |
| Synonym | Solvent Red 140 |
| InChI Key | OALHHIHQOFIMEF-UHFFFAOYSA-N |
| SMILES | OC1=C(I)C2=C(C=C1I)C1(OC(=O)C3=C1C=CC=C3)C1=CC(I)=C(O)C(I)=C1O2 |
| IUPAC Name | 3',6'-dihydroxy-2',4',5',7'-tetraiodo-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one |
| Molecular Weight (g/mol) | 835.90 |
| PubChem CID | 3259 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction