missing translation for 'onlineSavingsMsg'
Learn More

Erythrosin B, 95%, pure, spirit soluble, ACROS Organics™

Product Code. 10348140 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10348140

Brand: Thermo Scientific Acros 191120050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 15905-32-5
Assay Percent Range 95%
Molecular Formula C20H8I4O5
MDL Number MFCD00005044
Beilstein 19, 231
InChI Key OALHHIHQOFIMEF-UHFFFAOYSA-N
Infrared Spectrum Authentic
SMILES OC1=C(I)C2=C(C=C1I)C1(OC(=O)C3=C1C=CC=C3)C1=CC(I)=C(O)C(I)=C1O2
IUPAC Name 3',6'-dihydroxy-2',4',5',7'-tetraiodo-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Weight (g/mol) 835.90
PubChem CID 3259
Formula Weight 835.9
Grade Pure
Packaging Glass bottle
UV_Vis 94% min.
UV Spectrometry Complies
Color Orange to Orange/Red
Quantity 5 g
Physical Form Powder
Chemical Name or Material Erythrosin B, 95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.