missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erythropoietin R Antibody (016), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89615AF532
This item is not returnable.
View return policy
Description
Erythropoietin R Monoclonal specifically detects Erythropoietin R in Human samples. It is validated for Western Blot.
Specifications
| Erythropoietin R | |
| Monoclonal | |
| Alexa Fluor 532 | |
| EPO-R, erythropoietin receptor, MGC138358 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Erythropoietin R (NP_000112.1; Met1-Pro250). | |
| 0.1 mL | |
| Angiogenesis, Cancer, Immunology, Phospho Specific, Signal Transduction | |
| 2057 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| 16 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction