missing translation for 'onlineSavingsMsg'
Learn More

Erythropoietin/EPO Rabbit anti-Human, Clone: EPO/3793R, Novus Biologicals™

Product Code. 18376393 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376393 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376393 Supplier Novus Biologicals Supplier No. NBP30715320UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Erythropoietin/EPO
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EPO/3793R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 2-4 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Host Species Rabbit
Immunogen Recombinant fragment (around aa 28-162) of human Erythropoietin/EPO protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cytokine Research, HIF Target Genes, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 2056
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.