missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erythropoietin/EPO Antibody (EPO/3793R), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08895AF750
This item is not returnable.
View return policy
Description
Erythropoietin/EPO Monoclonal antibody specifically detects Erythropoietin/EPO in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Erythropoietin/EPO | |
| Monoclonal | |
| Alexa Fluor 750 | |
| EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
| Recombinant fragment (around aa 28-162) of human Erythropoietin/EPO protein (exact sequence is proprietary) (Uniprot: P01588) | |
| 100 μg | |
| Angiogenesis, Cancer, Cytokine Research, HIF Target Genes, Hypoxia | |
| 2056 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| EPO/3793R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction