missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erythropoietin/EPO Antibody (EPO/1367), Alexa Fluor™ 405, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Erythropoietin/EPO Monoclonal antibody specifically detects Erythropoietin/EPO in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
Specifications
| Antigen | Erythropoietin/EPO |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Classification | Monoclonal |
| Clone | EPO/1367 |
| Conjugate | Alexa Fluor 405 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 |
| Host Species | Mouse |
| Immunogen | Recombinant fragment (around aa 28-162) of human Erythropoietin/EPO protein (exact sequence is proprietary) (Uniprot: P01588) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?