missing translation for 'onlineSavingsMsg'
Learn More

Erythropoietin/EPO Antibody (971007) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30735629 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735629 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735629 Supplier Novus Biologicals Supplier No. FAB2873MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Erythropoietin/EPO Monoclonal antibody specifically detects Erythropoietin/EPO in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Erythropoietin/EPO
Applications Immunohistochemistry
Classification Monoclonal
Clone 971007
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human Erythropoietin/EPO protein, Ala28-Arg193, Accession # CAA26094
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cytokine Research, HIF Target Genes, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 2056
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.