missing translation for 'onlineSavingsMsg'
Learn More

Erythromycin (White Crystalline Powder), Fisher BioReagents

Product Code. 10583315
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10583315 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10583315 Supplier Fisher BioReagents Supplier No. BP92025

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Erythromycin is used as an inhibitor in cell-free protein biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Type Erythromycin
Physical Form Solid
CAS 114-07-8
pH 8 to 10.5
Melting Point 138°C
Ignition Residue 0.2% max.
Packaging Poly Bottle
Specific Rotation Inclusive between -78° (+ or -)
Color White
Quantity 25 g
Assay Percent Range 85%
Molecular Formula C37H67NO13
MDL Number MFCD00084654
Merck Index 15, 3739
Synonym E-Mycin, Erythrocin
InChI Key ULGZDMOVFRHVEP-RWJQBGPGSA-N
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O
IUPAC Name (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradecane-2,10-dione
Molecular Weight (g/mol) 733.94
PubChem CID 12560
ChEBI CHEBI:42355
Formula Weight 733.94
Chemical Name or Material Erythromycin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.