missing translation for 'onlineSavingsMsg'
Learn More

Erythromycin N-2-Propenyl Bromide, TRC

Product Code. 30380880
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380880 10 mg 10mg
30385443 50 mg 50mg
30373739 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380880 Supplier Toronto Research Chemicals Supplier No. TRCE64995510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Erythromycin N-2-Propenyl Bromide
Analyte or Component Names Erythromycin N-2-Propenyl Bromide
Shipping Condition Room Temperature
Molecular Formula C40H72NO13•Br
Synonym (3R,4S)-N-Allyl-2-(((3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-14-ethyl-7,12,13-trihydroxy-4-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-3,5,7,9,11,13-hexamethyl-2,10-dioxooxacyclotetradecan-6-yl)oxy)-3-hydroxy-N,N,6-trimethyltetrahydro-2H-pyran-4-aminium Bromide
InChI Formula InChI=1S/C40H72NO13.BrH/c1-15-17-41(12,13)27-18-22(4)50-37(31(27)43)54-35-24(6)32(53-29-20-39(10,49-14)34(45)26(8)51-29)25(7)36(46)52-28(16-2)40(11,48)33(44)23(5)30(42)21(3)19-38(35,9)47;/h15,21-29,31-35,37,43-45,47-48H,1,16-20H2,2-14H3;1H/q+1;/p-1/t21-,22?,23+,24+,25-,26+,27+,28-,29+,31-,32+,33-,34+,35-,37?,38-,39-,40-;/m1./s1
Solution Type Neat
SMILES C[C@@H]1C[C@](O)(C)[C@H](OC2[C@H](O)[C@@H]([N+](C)(CC=C)C)CC(C)O2)[C@@H](C)[C@H](O[C@@H]3O[C@@H](C)[C@H](O)[C@](C)(OC)C3)[C@@H](C)C(O[C@H](CC)[C@](O)(C)[C@H](O)[C@@H](C)C1=O)=O.[Br-]
IUPAC Name (3R,4S)-N-allyl-2-(((3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-14-ethyl-7,12,13-trihydroxy-4-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-3,5,7,9,11,13-hexamethyl-2,10-dioxooxacyclotetradecan-6-yl)oxy)-3-hydroxy-N,N,6-trimethyltetrahydro-2H-pyran-4-aminium bromide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 854.9
Quantity 10 mg
Formula Weight 853.42
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.