missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erythromycin N-2-Propenyl Bromide, TRC
Specifications
Specifications
| Chemical Name or Material | Erythromycin N-2-Propenyl Bromide |
| Analyte or Component Names | Erythromycin N-2-Propenyl Bromide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C40H72NO13•Br |
| Synonym | (3R,4S)-N-Allyl-2-(((3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-14-ethyl-7,12,13-trihydroxy-4-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-3,5,7,9,11,13-hexamethyl-2,10-dioxooxacyclotetradecan-6-yl)oxy)-3-hydroxy-N,N,6-trimethyltetrahydro-2H-pyran-4-aminium Bromide |
| InChI Formula | InChI=1S/C40H72NO13.BrH/c1-15-17-41(12,13)27-18-22(4)50-37(31(27)43)54-35-24(6)32(53-29-20-39(10,49-14)34(45)26(8)51-29)25(7)36(46)52-28(16-2)40(11,48)33(44)23(5)30(42)21(3)19-38(35,9)47;/h15,21-29,31-35,37,43-45,47-48H,1,16-20H2,2-14H3;1H/q+1;/p-1/t21-,22?,23+,24+,25-,26+,27+,28-,29+,31-,32+,33-,34+,35-,37?,38-,39-,40-;/m1./s1 |
| Solution Type | Neat |
| SMILES | C[C@@H]1C[C@](O)(C)[C@H](OC2[C@H](O)[C@@H]([N+](C)(CC=C)C)CC(C)O2)[C@@H](C)[C@H](O[C@@H]3O[C@@H](C)[C@H](O)[C@](C)(OC)C3)[C@@H](C)C(O[C@H](CC)[C@](O)(C)[C@H](O)[C@@H](C)C1=O)=O.[Br-] |
| IUPAC Name | (3R,4S)-N-allyl-2-(((3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-14-ethyl-7,12,13-trihydroxy-4-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-3,5,7,9,11,13-hexamethyl-2,10-dioxooxacyclotetradecan-6-yl)oxy)-3-hydroxy-N,N,6-trimethyltetrahydro-2H-pyran-4-aminium bromide |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?