missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erythromycin, ∽98%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
100 g
Unit Size:
100g
1g
5g
Description
- It Used in cell culture applications at approximately 100 mg/liter.
- Depending upon the strain of bacteria, erythromycin has been used between 50 and 200 mg/L to control bacterial growth.
- Erythromycin resistance can be induced in bacteria.
- Erythromycin is a macrolide antibiotic that bind to the 50S ribosomal subunit and inhibit protein synthesis.
- This is an antimicrobial substance produced by the growth of certain strains of Streptomyces erythreus.
- It is a mixture consisting largely of erythromycin A with lesser amounts of erythromycins B and C.
Specifications
Specifications
| Type | Erythromycin |
| Physical Form | Powder |
| Color | White |
| Quantity | 100 g |
| Assay Percent Range | ∽ 98% |
| Synonym | E-mycin, Erycin |
| Formula Weight | 733.95 |
| Chemical Name or Material | Erythromycin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction