missing translation for 'onlineSavingsMsg'
Learn More

Erythromycin, ∽98%, MP Biomedicals™

Product Code. 11431631 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
Unit Size:
100g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431631 5 g 5g
11421631 1 g 1g
11451631 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431631 Supplier MP Biomedicals Supplier No. 0219019705
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity antibiotic with>98% purity for research and laboratory applications.

  • It Used in cell culture applications at approximately 100 mg/liter.
  • Depending upon the strain of bacteria, erythromycin has been used between 50 and 200 mg/L to control bacterial growth.
  • Erythromycin resistance can be induced in bacteria.
  • Erythromycin is a macrolide antibiotic that bind to the 50S ribosomal subunit and inhibit protein synthesis.
  • This is an antimicrobial substance produced by the growth of certain strains of Streptomyces erythreus.
  • It is a mixture consisting largely of erythromycin A with lesser amounts of erythromycins B and C.

Chemical Identifiers

CAS 114-07-8
Molecular Formula C37H67NO13
Molecular Weight (g/mol) 733.94
MDL Number MFCD00084654
InChI Key ULGZDMOVFRHVEP-RWJQBGPGSA-N
PubChem CID 12560
ChEBI CHEBI:42355
IUPAC Name (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradecane-2,10-dione
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O

Specifications

Type Erythromycin
Physical Form Powder
Color White
Quantity 5 g
Assay Percent Range ∽ 98%
Synonym E-mycin, Erycin
Formula Weight 733.95
Percent Purity 98%
Grade Not Available
Chemical Name or Material Erythromycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.