missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (-)-Erythromycin, 98%

Product Code. 10338080 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10338080 5 g Plastic Bottle 5g
10053110 25 g Plastic Bottle 25g
10155000 100 g Plastic Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10338080 Supplier Thermo Scientific Chemicals Supplier No. 227330050
  • ONLINE EXCLUSIVE PRICE
    £62.40
    £54.65 / 5g
    Save £7.75
    12% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Erythromycin (E-Mycin or erythrocin), CAS # 114-07-8, is a broad-spectrum antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Erythromycin is a macrolide antibiotic that works by binding the 50S ribosomal subunit and inhibiting polypeptide formation

• Erythromycin was isolated from Saccharopolyspora erythraea

Applications

• Erythromycin can be used to disrupt the growth of gram-positive and gram-negative bacteria

• Erythromycin has been shown to have motor effects in the gastrointestinal system and can be used for laboratory research on migrating motor complex activity

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114-07-8
Molecular Formula C37H67NO13
Molecular Weight (g/mol) 733.94
MDL Number MFCD00084654
InChI Key ULGZDMOVFRHVEP-RWJQBGPGSA-N
PubChem CID 12560
ChEBI CHEBI:42355
IUPAC Name (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-7,12,13-trihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dione
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O

Specifications

Type (-)-Erythromycin
Physical Form Powder
Melting Point 138.0°C to 140.0°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Specific Rotation −74.50° (20°C c=2,ethanol)
Water 6.5% max. (K.F.)
Color White to Yellow
Quantity 5 g
Assay Percent Range 98%
Merck Index 15, 3739
Solubility Information Solubility in water: +/- 2 mg/mg. Other solubilities: freely soluble in alcohols, acetone, chloroform,, acetonitrile, ethyl acetate, moderately soluble in ether, ethylenedichloride, and amyl acetate
Formula Weight 733.94
Percent Purity 98%
Chemical Name or Material (-)-Erythromycin, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.