missing translation for 'onlineSavingsMsg'
Learn More

Erythrocyte Specific Antibody (SFL23.6), Novus Biologicals™

Product Code. 18427351 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
Unit Size:
0.02mg
0.10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427351 0.02 mg 0.02mg
18470662 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18427351 Supplier Novus Biologicals Supplier No. NBP2328450.02mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Erythrocyte Specific Monoclonal antibody specifically detects Erythrocyte Specific in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Erythrocyte Specific
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone SFL23.6
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Alias erythroid cell surface antigen
Host Species Mouse
Immunogen Hepatocytes from a 20-22 week-old human fetus
Purification Method Protein A or G purified
Quantity 0.02 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.