missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-erythro-Aleuritic acid, 95%

Product Code. 11318225
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318225 5 g 5g
11328225 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318225 Supplier Thermo Scientific Alfa Aesar Supplier No. L08542.06
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 533-87-9 | C16H32O5 | 304.427 g/mol

α-Aleuritic acid is a major ingredient of shellac and is also a starting reagent in the manufacture of perfumes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-Aleuritic acid is a major ingredient of shellac and is also a starting reagent in the manufacture of perfumes.

Solubility
Soluble in water, alcohol, acetic acid and acetone.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-87-9
Molecular Formula C16H32O5
Molecular Weight (g/mol) 304.427
MDL Number MFCD00135596
InChI Key MEHUJCGAYMDLEL-LSDHHAIUSA-N
Synonym aleuretic acid, 8,9,15-trihydroxypentadecane-1-carboxylic acid, 9,10,16-trihydroxypalmitic acid, 10r,9s-9,10,16-trihydroxyhexadecanoic acid, 9s,10r-9,10,16-trihydroxyhexadecanoic acid, unii-442lq2k03a component
PubChem CID 7269316
IUPAC Name (9S,10R)-9,10,16-trihydroxyhexadecanoic acid
SMILES C(CCCC(C(CCCCCCO)O)O)CCCC(=O)O

Specifications

Melting Point 97°C to 101°C
Odor Odorless
Quantity 5 g
Beilstein 1727724
Merck Index 14,231
Solubility Information Soluble in water,alcohol,acetic acid and acetone.
Formula Weight 304.43
Percent Purity 95%
Chemical Name or Material (+/-)-erythro-Aleuritic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.