missing translation for 'onlineSavingsMsg'
Learn More

Erythritol, TRC

Product Code. 30372126
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372126 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372126 Supplier Toronto Research Chemicals Supplier No. TRCE65010050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material meso-Erythritol
CAS 149-32-6
Purity Grade Notes HPLC
Molecular Formula C4 H10 O4
Quantity 50 mg
Synonym Erythritol, (2R,3S)-Butane-1,2,3,4-tetrol, meso-erythritol, 1,2,3,4-Butanetetrol, (2R,3S)-rel-, 1,2,3,4-Butanetetrol, (R*,S*)-, Erythritol (8CI), (2R,3S)-Butane-1,2,3,4-tetraol, C* Eridex 16954, C*Eridex, Cargill Zerose 16957, Eridex F 16960, Erythrit, Erythritol 100M, Erythrol, F 8015, Lakanto-S, Mesoerythritol, NIK 242, NSC 8099, Phycitol, Zerose, Zerose 01657, Zerose 16952, Zerose Tm 16957, meso-Erythritol
InChI Formula InChI=1S/C4H10O4/c5-1-3(7)4(8)2-6/h3-8H,1-2H2/t3-,4+
SMILES OC[C@@H](O)[C@@H](O)CO
IUPAC Name (2S,3R)-butane-1,2,3,4-tetrol
Molecular Weight (g/mol) 122.12
Formula Weight 122.0579
Percent Purity >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.