missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erythritol, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Erythritol
Specifications
Specifications
| Chemical Name or Material | meso-Erythritol |
| CAS | 149-32-6 |
| Analyte or Component Names | meso-Erythritol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C4 H10 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Erythritol, (2R,3S)-Butane-1,2,3,4-tetrol, meso-erythritol, 1,2,3,4-Butanetetrol, (2R,3S)-rel-, 1,2,3,4-Butanetetrol, (R*,S*)-, Erythritol (8CI), (2R,3S)-Butane-1,2,3,4-tetraol, C* Eridex 16954, C*Eridex, Cargill Zerose 16957, Eridex F 16960, Erythrit, Erythritol 100M, Erythrol, F 8015, Lakanto-S, Mesoerythritol, NIK 242, NSC 8099, Phycitol, Zerose, Zerose 01657, Zerose 16952, Zerose Tm 16957, meso-Erythritol |
| InChI Formula | InChI=1S/C4H10O4/c5-1-3(7)4(8)2-6/h3-8H,1-2H2/t3-,4+ |
| SMILES | OC[C@@H](O)[C@@H](O)CO |
| IUPAC Name | (2S,3R)-butane-1,2,3,4-tetrol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction