missing translation for 'onlineSavingsMsg'
Learn More

Erythralosamine, TRC

Product Code. 30382877
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382877 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382877 Supplier Toronto Research Chemicals Supplier No. TRCE65850525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Erythralosamine
CAS 546-57-6
Molecular Formula C29 H49 N O8
Quantity 25 mg
Synonym Erythromycin, 3-O-de(2,6-dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)-10,11-didehydro-9-deoxo-6,11,12-trideoxy-6,9:9,12-diepoxy- (9CI), Erythralosamine (6CI,7CI,8CI), 6,15,16-Trioxatricyclo[10.2.1.11,4]hexadecane, erythromycin deriv., (1S,4S,5R,8R,9S,10S,11R,12R,14R)-5-Ethyl-9-hydroxy-2,4,8,10,12,14-hexamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-6,15,16-trioxatricyclo[10.2.1.11,4]hexadec-2-en-7-one, Erythralosamine
InChI Formula InChI=1S/C29H49NO8/c1-11-21-27(7)13-15(2)29(37-27)16(3)14-28(8,38-29)24(18(5)22(31)19(6)25(33)35-21)36-26-23(32)20(30(9)10)12-17(4)34-26/h13,16-24,26,31-32H,11-12,14H2,1-10H3/t16-,17-,18+,19-,20+,21-,22+,23-,24-,26+,27+,28-,29?/m1/s1
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@@]3(C)C[C@@H](C)C4(O3)O[C@@]1(C)C=C4C
Molecular Weight (g/mol) 539.7
Formula Weight 539.3458
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.