missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERVMER34-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552836
This item is not returnable.
View return policy
Description
Immunogen sequence: GNGVYKGFPP KWSGRCGLGY LVPSLTRYLT LNASQITNLR SFIHKVTPHR CTQGDTDNPP LYCNPKDNST IRALFPSLGT YDLEKAILNI SKAMEQEFSA TKQTLEAHQS KVSSLASASR KDHVLDIPTT QRQTACGTVG K Highest antigen sequence identity to the following orthologs: Mouse - 23%, Rat - 20%.
Specifications
| ERVMER34-1 | |
| Polyclonal | |
| Unconjugated | |
| ERVMER34-1 | |
| Endogenous retroviral envelope protein HEMO; Endogenous retroviral envelope protein HEMO, 48 kDa form; Endogenous retroviral envelope protein HEMO, secreted form; endogenous retrovirus group MER34 member 1; endogenous retrovirus group MER34 member 1 Env polyprotein; endogenous retrovirus group MER34, member 1; ERVMER34-1; HEMO; HERV-MER_4q12 provirus ancestral Env polyprotein; Human endogenous MER34 (medium-reiteration-frequency-family-34) open reading frame; Human endogenous MER34 ORF; LP9056; LP9056 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100288413 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H9K5 | |
| ERVMER34-1 | |
| Recombinant protein corresponding to Human ERVMER34-1. Recombinant protein control fragment (Product #RP-90380). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction