missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERV3-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £470.00
Specifications
| Antigen | ERV3-1 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18685617
|
Novus Biologicals
NBP2-62602 |
100 μg |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610249
|
Novus Biologicals
NBP2-62602-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
ERV3-1 Polyclonal antibody specifically detects ERV3-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Spezifikation
| ERV3-1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Endogenous Retroviral Sequence 3, Endogenous Retroviral Sequence 3 (Includes Zinc Finger Protein H-Plk/HPF9), Endogenous Retrovirus Group 3 Member 1, Endogenous Retrovirus Group 3 Member 1 Env Polyprotein, Endogenous Retrovirus Group 3 Member 1, Envelope, Endogenous Retrovirus Group 3, Member 1, envelope polyprotein, ERV3, ERV3 Envelope Protein, ERV-3 Envelope Protein, ERV3-1 Envelope Protein, ERV-R, ERV-R Envelope Protein, HERVR, HERV-R, HERV-R_7q21.2 Provirus Ancestral Env Polyprotein | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QLAENIASSLHVASCYVCGGMNMGDQWPWEARELMPQDNFTLTASSLEPAPSSQSIWFLKTSIIGKFCIARWGKAFTDPVGELTCLGQQYYNETLGKTLWRGKSNNSESPHPSPFSRFPSLNHSWYQLEA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 2086 | |
| IgG | |
| Protein A purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts