missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERV3-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68667
This item is not returnable.
View return policy
Description
ERV3-1 Polyclonal antibody specifically detects ERV3-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ERV3-1 | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Endogenous Retroviral Sequence 3, Endogenous Retroviral Sequence 3 (Includes Zinc Finger Protein H-Plk/HPF9), Endogenous Retrovirus Group 3 Member 1, Endogenous Retrovirus Group 3 Member 1 Env Polyprotein, Endogenous Retrovirus Group 3 Member 1, Envelope, Endogenous Retrovirus Group 3, Member 1, envelope polyprotein, ERV3, ERV3 Envelope Protein, ERV-3 Envelope Protein, ERV3-1 Envelope Protein, ERV-R, ERV-R Envelope Protein, HERVR, HERV-R, HERV-R_7q21.2 Provirus Ancestral Env Polyprotein | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VCYDPKSSPGIWFEIHVGSKEGDLLNQTKVFPSGKDVVSLYFDVCQIVSMGSLFPVIFSSMEYYSSCHKNRYAHPACSTDSPVTTCWDCTTWSTNQQSLGPIMLTKIPLEPDCKTSTCNSVNLTILEPDQPIWTTGLKAPL | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 2086 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction