missing translation for 'onlineSavingsMsg'
Learn More

Erucic Acid, TRC

Product Code. 17981675
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17981675 10 g 10g
17933075 5 g 5g
17972865 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17981675 Supplier Toronto Research Chemicals Supplier No. TRCE64990010G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-86-7
Molecular Formula C22 H42 O2
Molecular Weight (g/mol) 338.57
Synonym 13-Docosenoic acid, (Z)- (8CI), (13Z)-13-Docosenoic acid, (13Z)-Docosenoic acid, (Z)-13-Docosenoic acid, (Z)-Erucic acid, 13(Z)-Docosenoic acid, 13-cis-Docosenoic acid, Erucic acid, Hystrene 2290, Jaric 22:1, Nouracid RE 07, Prifrac 2990, cis-13-Docosenoic acid, cis-13-Erucic acid, δ13-cis-Docosenoic acid, Erucic Acid
IUPAC Name (Z)-docos-13-enoic acid
SMILES CCCCCCCC\C=C/CCCCCCCCCCCC(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C22H42O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h9-10H,2-8,11-21H2,1H3,(H,23,24)/b10-9-
Formula Weight 338.3185
Chemical Name or Material Erucic Acid
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.