missing translation for 'onlineSavingsMsg'
Learn More

Erucic Acid, TRC

Product Code. 17933075
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17933075 5 g 5g
17981675 10 g 10g
17972865 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17933075 Supplier Toronto Research Chemicals Supplier No. TRCE6499005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-86-7
Molecular Formula C22 H42 O2
Molecular Weight (g/mol) 338.57
Synonym 13-Docosenoic acid, (Z)- (8CI), (13Z)-13-Docosenoic acid, (13Z)-Docosenoic acid, (Z)-13-Docosenoic acid, (Z)-Erucic acid, 13(Z)-Docosenoic acid, 13-cis-Docosenoic acid, Erucic acid, Hystrene 2290, Jaric 22:1, Nouracid RE 07, Prifrac 2990, cis-13-Docosenoic acid, cis-13-Erucic acid, δ13-cis-Docosenoic acid, Erucic Acid
IUPAC Name (Z)-docos-13-enoic acid
SMILES CCCCCCCC\C=C/CCCCCCCCCCCC(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C22H42O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h9-10H,2-8,11-21H2,1H3,(H,23,24)/b10-9-
Formula Weight 338.3185
Chemical Name or Material Erucic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.