missing translation for 'onlineSavingsMsg'
Learn More

Ertapenem N-Isobutoxycarbonyl O-(4-Nitrobenzyl), TRC

Product Code. 30387800
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
2.5 mg
Unit Size:
0.25mg
1mg
2.50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387800 2.5 mg 2.50mg
30384398 1 mg 1mg
30387434 0.25 mg 0.25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387800 Supplier Toronto Research Chemicals Supplier No. TRCE6350652.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ertapenem N-Isobutoxycarbonyl O-(4-Nitrobenzyl)
Analyte or Component Names Ertapenem N-Isobutoxycarbonyl O-(4-Nitrobenzyl)
Shipping Condition Room Temperature
Molecular Formula C35 H40 N4 O11 S
Synonym (4R,5S,6S)-4-Nitrobenzyl 6-((R)-1-Hydroxyethyl)-3-(((3S,5S)-1-(isobutoxycarbonyl)-5-((3-(methoxycarbonyl)phenyl)carbamoyl)pyrrolidin-3-yl)thio)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate
InChI Formula InChI=1S/C35H40N4O11S/c1-18(2)16-50-35(45)37-15-25(14-26(37)31(41)36-23-8-6-7-22(13-23)33(43)48-5)51-30-19(3)28-27(20(4)40)32(42)38(28)29(30)34(44)49-17-21-9-11-24(12-10-21)39(46)47/h6-13,18-20,25-28,40H,14-17H2,1-5H3,(H,36,41)/t19-,20-,25+,26-,27-,28-/m1/s1
Solution Type Neat
SMILES COC(=O)c1cccc(NC(=O)[C@H]2C[C@@H](CN2C(=O)OCC(C)C)SC3=C(N4[C@H]([C@H]3C)[C@@H]([C@@H](C)O)C4=O)C(=O)OCc5ccc(cc5)[N+](=O)[O-])c1
IUPAC Name (4-nitrophenyl)methyl (4R,5S,6S)-6-[(1R)-1-hydroxyethyl]-3-[(3S,5R)-5-[(3-methoxycarbonylphenyl)carbamoyl]-1-(2-methylpropoxycarbonyl)pyrrolidin-3-yl]sulfanyl-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 724.777
Quantity 2.5 mg
Formula Weight 724.241
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.