missing translation for 'onlineSavingsMsg'
Learn More

Ertapenem N-Carboxybenzyl Dimethyl Ester, TRC

Product Code. 30384289
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
2.5 mg
Unit Size:
0.25mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384289 1 mg 1mg
30384028 2.5 mg 2.5mg
30382074 0.25 mg 0.25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384289 Supplier Toronto Research Chemicals Supplier No. TRCE6350551MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ertapenem N-Carboxybenzyl Dimethyl Ester
Analyte or Component Names Ertapenem N-Carboxybenzyl Dimethyl Ester
Shipping Condition Room Temperature
Molecular Formula C32 H35 N3 O9 S
Synonym (4R,5S,6S)-3-(((3S,5S)-1-((Benzyloxy)carbonyl)-5-((3-(methoxycarbonyl)phenyl)carbamoyl)pyrrolidin-3-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate Sodium Salt
InChI Formula InChI=1S/C32H35N3O9S/c1-17-25-24(18(2)36)29(38)35(25)26(31(40)43-4)27(17)45-22-14-23(28(37)33-21-12-8-11-20(13-21)30(39)42-3)34(15-22)32(41)44-16-19-9-6-5-7-10-19/h5-13,17-18,22-25,36H,14-16H2,1-4H3,(H,33,37)/t17-,18-,22-,23-,24-,25-/m1/s1
Solution Type Neat
SMILES COC(=O)C1=C(S[C@@H]2C[C@@H](N(C2)C(=O)OCc3ccccc3)C(=O)Nc4cccc(c4)C(=O)OC)[C@H](C)[C@@H]5[C@@H]([C@@H](C)O)C(=O)N15
IUPAC Name methyl (4R,5S,6S)-6-[(1R)-1-hydroxyethyl]-3-[(3R,5R)-5-[(3-methoxycarbonylphenyl)carbamoyl]-1-phenylmethoxycarbonylpyrrolidin-3-yl]sulfanyl-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 637.7
Quantity 1 mg
Formula Weight 637.209
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.