missing translation for 'onlineSavingsMsg'
Learn More

Ertapenem Disodium 90%, TRC

Product Code. 30381113
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381113 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381113 Supplier Toronto Research Chemicals Supplier No. TRCE635000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ertapenem Disodium
CAS 153832-38-3
Molecular Formula C22 H23 N3 O7 S . 2 Na
Quantity 100 mg
Synonym (4R,5S,6S)-3-[[(3S,5S)-5-[[(3-carboxyphenyl)amino]carbonyl]-3-pyrrolidinyl]thio]-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-Azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid sodium salt (1:2), 1-Azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, 3-[[(3S,5S)-5-[[(3-carboxyphenyl)amino]carbonyl]-3-pyrrolidinyl]thio]-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-, disodium salt, (4R,5S,6S)- (9CI), 1-Azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid, 3-[[5-[[(3-carboxyphenyl)amino]carbonyl]-3-pyrrolidinyl]thio]-6-(1-hydroxyethyl)-4-methyl-7-oxo-, disodium salt, [4R-[3(3S*,5S*),4α,5β,6β(R*)]]-, Ertapenem disodium, L 749
InChI Formula InChI=1S/C22H25N3O7S.2Na/c1-9-16-15(10(2)26)20(28)25(16)17(22(31)32)18(9)33-13-7-14(23-8-13)19(27)24-12-5-3-4-11(6-12)21(29)30;;/h3-6,9-10,13-16,23,26H,7-8H2,1-2H3,(H,24,27)(H,29,30)(H,31,32);;/q;2*+1/p-2/t9-,10-,13+,14+,15-,16-;;/m1../s1
SMILES [Na+].[Na+].C[C@@H](O)[C@@H]1[C@H]2[C@@H](C)C(=C(N2C1=O)C(=O)[O-])S[C@@H]3CN[C@@H](C3)C(=O)Nc4cccc(c4)C(=O)[O-]
IUPAC Name disodium;(4R,5S,6S)-3-[(3S,5S)-5-[(3-carboxylatophenyl)carbamoyl]pyrrolidin-3-yl]sulfanyl-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate
Molecular Weight (g/mol) 519.48
Formula Weight 519.1052
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.