missing translation for 'onlineSavingsMsg'
Learn More

ERR gamma/NR3B3 Rabbit anti-Human, Mouse, Rat, Clone: 2W3A9, Novus Biologicals™

Product Code. 18372077 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372077 100 μg 100µL
18369006 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18372077 Supplier Novus Biologicals Supplier No. NBP316836100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ERR gamma/NR3B3 Monoclonal antibody specifically detects ERR gamma/NR3B3 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERR gamma/NR3B3
Applications Western Blot
Classification Monoclonal
Clone 2W3A9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DKFZp781L1617, ERR gamma-2, ERR3, ERRG2, ERRgamma, Estrogen receptor-related protein 3, estrogen-related receptor gamma, FLJ16023, KIAA0832, NR3B3, Nuclear receptor subfamily 3 group B member 3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ERR gamma/NR3B3 (P62508).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2104
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.