missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERR gamma/NR3B3 Antibody (OTI1E5), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70691B
This item is not returnable.
View return policy
Description
ERR gamma/NR3B3 Monoclonal specifically detects ERR gamma/NR3B3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ERR gamma/NR3B3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| ESRRG | |
| Recombinant protein fragment corresponding to amino acids 24-263 of human ERR gamma/NR3B3 (NP_001429) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1E5 | |
| Western Blot, Immunohistochemistry | |
| DKFZp781L1617, ERR gamma-2, ERR3, ERRG2, ERRgamma, Estrogen receptor-related protein 3, estrogen-related receptor gamma, FLJ16023, KIAA0832, NR3B3, Nuclear receptor subfamily 3 group B member 3 | |
| Mouse | |
| Affinity Purified | |
| GPCR | |
| 2104.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction