missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERR gamma/NR3B3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-00182
This item is not returnable.
View return policy
Description
ERR gamma/NR3B3 Polyclonal specifically detects ERR gamma/NR3B3 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| ERR gamma/NR3B3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| DKFZp781L1617, ERR gamma-2, ERR3, ERRG2, ERRgamma, Estrogen receptor-related protein 3, estrogen-related receptor gamma, FLJ16023, KIAA0832, NR3B3, Nuclear receptor subfamily 3 group B member 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize the reported isoform NP_001429.2. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.2 - 0.6 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_001429.2 | |
| ESRRG | |
| Peptide with sequence CRMSNKDRHIDS corresponding to internal region according to NP_001429.2, NP_996317.1, NP_996318.1. | |
| 0.1 mg | |
| GPCR | |
| 2104 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction