missing translation for 'onlineSavingsMsg'
Learn More

ERR alpha/NR3B1 Rabbit anti-Human, Mouse, Clone: 2R10S3, Novus Biologicals™

Product Code. 18322705 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322705 100 μg 100µL
18330656 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18322705 Supplier Novus Biologicals Supplier No. NBP316349100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ERR alpha/NR3B1 Monoclonal antibody specifically detects ERR alpha/NR3B1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERR alpha/NR3B1
Applications Western Blot
Classification Monoclonal
Clone 2R10S3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ERR1Estrogen receptor-like 1, ERRa, ESRL1ERR-alpha, estrogen-related receptor alphaERRalpha, NR3B1Nuclear receptor subfamily 3 group B member 1, steroid hormone receptor ERR1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ERR alpha/NR3B1 (P11474).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2101
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.