missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERR alpha/NR3B1 Antibody (6F1E6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61768
This item is not returnable.
View return policy
Description
ERR alpha/NR3B1 Monoclonal specifically detects ERR alpha/NR3B1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| ERR alpha/NR3B1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| ERR1Estrogen receptor-like 1, ERRa, ESRL1ERR-alpha, estrogen-related receptor alphaERRalpha, NR3B1Nuclear receptor subfamily 3 group B member 1, steroid hormone receptor ERR1 | |
| Mouse | |
| 66.2 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 6F1E6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ESRRA | |
| Purified recombinant fragment of human ESRRA (AA: 198-376) expressed in E. Coli. | |
| Protein G purified | |
| Cancer, Cell Biology, Cell Cycle and Replication, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| 2101 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction