missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERp72 Antibody (OTI2B9), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71488AF488
This item is not returnable.
View return policy
Description
ERp72 Monoclonal specifically detects ERp72 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ERp72 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PDIA4 | |
| Full length recombinant protein of human ERp72 (NP_004902) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2B9 | |
| Western Blot, Immunohistochemistry | |
| Endoplasmic reticulum resident protein 70, Endoplasmic reticulum resident protein 72, ER protein 70, ERp70, ERP70ER protein 72, ERp72, ERp-72, ERP72EC 5.3.4.1, intestinal-related), protein disulfide isomerase family A, member 4, protein disulfide isomerase related protein (calcium-binding protein, protein disulfide isomerase-associated 4, protein disulfide-isomerase A4 | |
| Mouse | |
| Affinity Purified | |
| Core ESC Like Genes, Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 9601.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction