missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERp57/PDIA3 Mouse anti-Bovine, Canine, Guinea Pig, Hamster, Human, Monkey, Mouse, Porcine, Rabbit, Rat, Clone: Map.ERp57 (GRP58), Novus Biologicals™
Mouse Monoclonal Antibody
£427.00 - £449.00
Specifications
| Antigen | ERp57/PDIA3 |
|---|---|
| Clone | Map.ERp57 (GRP58) |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18286941
|
Novus Biologicals
NBP2-59689-50UG |
50 μg |
£449.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18259293
|
Novus Biologicals
NBP2-59689-200UG |
200 μg |
£427.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ERp57/PDIA3 Monoclonal specifically detects ERp57/PDIA3 in Human, Mouse, Rat, Porcine, Bovine, Canine, Guinea Pig, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| ERp57/PDIA3 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, ER Markers, Membrane Trafficking and Chaperones | |
| PBS pH7.4, 50% glycerol with 0.09% Sodium Azide | |
| 2923 | |
| Human recombinant ERp57 (Grp58) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Map.ERp57 (GRP58) | |
| Western Blot 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Guinea Pig, Hamster, Monkey, Rabbit | |
| Disulfide isomerase ER-60, EC 5.3.4.1, endoplasmic reticulum P58, Endoplasmic reticulum resident protein 57, Endoplasmic reticulum resident protein 60, ER protein 60, ER60, ERP57, ERp5758 kDa glucose-regulated protein, ERP60, ERp6058 kDa microsomal protein, ERp61, glucose regulated protein, 58kDa, GRP57, GRP58ER protein 57, HsT17083, P58, phospholipase C-alpha, PI-PLC, protein disulfide isomerase family A, member 3, protein disulfide isomerase-associated 3, protein disulfide-isomerase A3 | |
| PDIA3 | |
| IgG1 | |
| Protein G purified | |
| 57 kDa |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title