missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERp57/PDIA3 Antibody (Map.ERp57 (GRP58)), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59689AF647
This item is not returnable.
View return policy
Description
ERp57/PDIA3 Monoclonal specifically detects ERp57/PDIA3 in Human, Mouse, Rat, Porcine, Bovine, Canine, Guinea Pig, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ERp57/PDIA3 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PDIA3 | |
| Human recombinant ERp57 (Grp58) | |
| 200 μL | |
| Cellular Markers, ER Markers, Membrane Trafficking and Chaperones | |
| 2923 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Guinea Pig, Hamster, Monkey, Rabbit | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Map.ERp57 (GRP58) | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Disulfide isomerase ER-60, EC 5.3.4.1, endoplasmic reticulum P58, Endoplasmic reticulum resident protein 57, Endoplasmic reticulum resident protein 60, ER protein 60, ER60, ERP57, ERp5758 kDa glucose-regulated protein, ERP60, ERp6058 kDa microsomal protein, ERp61, glucose regulated protein, 58kDa, GRP57, GRP58ER protein 57, HsT17083, P58, phospholipase C-alpha, PI-PLC, protein disulfide isomerase family A, member 3, protein disulfide isomerase-associated 3, protein disulfide-isomerase A3 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects approx 40kDa. Binds specifically to the N-terminus of Rhodopsin. Does not detect Rhodopsin in invertebrates. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction