missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERp57/PDIA3 Antibody (4F9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59695-25ug
This item is not returnable.
View return policy
Description
ERp57/PDIA3 Monoclonal specifically detects ERp57/PDIA3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ERp57/PDIA3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| Disulfide isomerase ER-60, EC 5.3.4.1, endoplasmic reticulum P58, Endoplasmic reticulum resident protein 57, Endoplasmic reticulum resident protein 60, ER protein 60, ER60, ERP57, ERp5758 kDa glucose-regulated protein, ERP60, ERp6058 kDa microsomal protein, ERp61, glucose regulated protein, 58kDa, GRP57, GRP58ER protein 57, HsT17083, P58, phospholipase C-alpha, PI-PLC, protein disulfide isomerase family A, member 3, protein disulfide isomerase-associated 3, protein disulfide-isomerase A3 | |
| Mouse | |
| 57 kDa | |
| 25 μg | |
| Cellular Markers, ER Markers, Membrane Trafficking and Chaperones | |
| 2923 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 4F9 | |
| Unconjugated | |
| PBS (pH 7.2), 50% glycerol with 0.09% Sodium Azide | |
| PDIA3 | |
| Human recombinant ERp57 (Grp58) protein corresponding to amino acids 25 - 505 (his-tagged) | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects approx 110kDa. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction