missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERp57 Monoclonal Antibody (MaP.Erp57)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA190869
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa Heat Shocked Cell Lysate.
This gene encodes a protein of the endoplasmic reticulum that interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesized glycoproteins. The protein was once thought to be a phospholipase; however, it has been demonstrated that the protein actually has protein disulfide isomerase activity. It is thought that complexes of lectins and this protein mediate protein folding by promoting formation of disulfide bonds in their glycoprotein substrates.
Specifications
| ERp57 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.05mM PMSF and 0.09% sodium azide; pH 7.2 | |
| P27773, P30101 | |
| PDIA3 | |
| Recombinant full length protein (Human). | |
| 50 μg | |
| Primary | |
| Human, Mouse, Canine, Monkey, Hamster, Pig | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| MaP.Erp57 | |
| Unconjugated | |
| PDIA3 | |
| 58 kDa glucose-regulated protein; 58 kDa microsomal protein; 58kDa; Disulfide isomerase ER-60; endoplasmic reticulum P58; endoplasmic reticulum protein; endoplasmic reticulum resident protein 57; Endoplasmic reticulum resident protein 60; epididymis secretory protein Li 269; epididymis secretory sperm binding protein Li 93n; ER p57; ER protein 57; ER protein 60; ER60; ER-60 protease; Erp; ERp 57; ERp57; ERP57 protein; ERP60; ERp61; glucose regulated protein 58; glucose regulated protein 58kD; glucose regulated protein, 58 kDa; glucose regulated protein, 58kDa; GRP 57; GRP 58; GRP57; Grp58; grp-58; HEL-S-269; HEL-S-93n; HIP-70; HsT17083; I79_003765; oxidoreductase ERp57; p58; PDI; PDIA 3; PDIA3; PDI-Q2; phospholipase C, alpha; phospholipase C-alpha; PI PLC; PI-PLC; PLC alpha; PLC[a]; Plca; protein disulfide isomerase associated 3; protein disulfide isomerase family A member 3; protein disulfide isomerase family A, member 3; protein disulfide isomerase-associated 3; protein disulfide-isomerase A3; Q-2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100156204, 100689343, 14827, 2923, 478279 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction