missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ERP44 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112156
This item is not returnable.
View return policy
Description
This product can be used in direct ELISA.
Mediates thiol-dependent retention in the early secretory pathway, forming mixed disulfides with substrate proteins through its conserved CRFS motif. Inhibits the calcium channel activity of ITPR1. May have a role in the control of oxidative protein folding in the endoplasmic reticulum. Required to retain ERO1L and ERO1LB in the endoplasmic reticulum.
Specifications
| ERP44 | |
| Polyclonal | |
| Unconjugated | |
| ERP44 | |
| 1110001E24Rik; AI849526; AL033348; BWK4; endoplasmic reticulum protein 44; endoplasmic reticulum resident protein 44; endoplasmic reticulum resident protein 44 kDa; endoplasmic reticulum resident protein 44kDa; endoplasmic reticulum resident protein ERp44; ER protein 44; Erp44; KIAA0573; OTTHUMP00000063799; PDIA10; protein disulfide isomerase family A, member 10; thioredoxin domain containing 4 (endoplasmic reticulum); thioredoxin domain-containing protein 4; Txndc4; UNQ532/PRO1075 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 23071 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9BS26 | |
| ERP44 | |
| Recombinant Human ERp44/TXNDC4 Protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction