Learn More
Invitrogen™ ERP44 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114141
Description
Immunogen sequence: LHREFHHGPD PTDTAPGEQA QDVASSPPES SFQKLAPSEY RYTLLRDRDE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 86%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 79%.
Mediates thiol-dependent retention in the early secretory pathway, forming mixed disulfides with substrate proteins through its conserved CRFS motif. Inhibits the calcium channel activity of ITPR1. May have a role in the control of oxidative protein folding in the endoplasmic reticulum. Required to retain ERO1L and ERO1LB in the endoplasmic reticulum.
Specifications
| ERP44 | |
| Polyclonal | |
| Unconjugated | |
| ERP44 | |
| 1110001E24Rik; AI849526; AL033348; BWK4; endoplasmic reticulum protein 44; endoplasmic reticulum resident protein 44; endoplasmic reticulum resident protein 44 kDa; endoplasmic reticulum resident protein 44kDa; endoplasmic reticulum resident protein ERp44; ER protein 44; Erp44; KIAA0573; OTTHUMP00000063799; PDIA10; protein disulfide isomerase family A, member 10; thioredoxin domain containing 4 (endoplasmic reticulum); thioredoxin domain-containing protein 4; Txndc4; UNQ532/PRO1075 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23071 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BS26 | |
| ERP44 | |
| Synthetic peptide directed towards the middle region of human TXNDC4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.