missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERP44 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38101-100ul
This item is not returnable.
View return policy
Description
ERP44 Polyclonal antibody specifically detects ERP44 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| ERP44 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| endoplasmic reticulum protein 44, endoplasmic reticulum resident protein 44 kDa, ER protein 44, ERp44, KIAA0573endoplasmic reticulum resident protein 44, PDIA10, protein disulfide isomerase family A, member 10, thioredoxin domain containing 4 (endoplasmic reticulum), Thioredoxin domain-containing protein 4, TXNDC4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 57-406 of human ERP44 (NP_055866.1).,, Sequence:, WCRFSQMLHPIFEEASDVIKEEFPNENQVVFARVDCDQHSDIAQRYRISKYPTLKLFRNGMMMKREYRGQRSVKALADYIRQQKSDPIQEIRDLAEITTLDRSKRNIIGYFEQKDSDNYRVFERVANILHDDCAFLSAFGDVSKPERYSGDNIIYKPPGHSAPDMVYLGAMTNFDVTYNWIQDKCVPLVREITFENGEELTEEGLPFLILFHMKEDTESLEIFQNEVARQLISEKGTINFLHADCDKFRHPLLHIQKTPADCPVIAIDSFRHMYVFGDFKDVLIPGKLKQFVFDLHSGKLHREFHHGPDPTDTAPGEQAQDVASSPPESSFQKLAPSEYRYTLLRDRDEL | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23071 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction