missing translation for 'onlineSavingsMsg'
Learn More

ERP44 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18430409 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430409 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430409 Supplier Novus Biologicals Supplier No. NBP297000MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ERP44 Polyclonal antibody specifically detects ERP44 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERP44
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias endoplasmic reticulum protein 44, endoplasmic reticulum resident protein 44 kDa, ER protein 44, ERp44, KIAA0573endoplasmic reticulum resident protein 44, PDIA10, protein disulfide isomerase family A, member 10, thioredoxin domain containing 4 (endoplasmic reticulum), Thioredoxin domain-containing protein 4, TXNDC4
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human ERP44 (Accession#: NP_055866.1; Met1-Asp402)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23071
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.